accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3PH3CPCH3H3C

CAS RN 64065-08-3

IUPAC Name: [(dimethylphosphanyl)methyl]dimethylphosphane
Synonyms: [(dimethylphosphanyl)methyl]dimethylphosphane
Molecular Weight (g/mol): 136.12
Molecular Formula: C5H14P2
InChi Key: MRNJHNUEBDGNEL-UHFFFAOYSA-N
SMILES: CP(C)CP(C)C
Boiling Point 
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)