accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6408-51-1

IUPAC Name: 5-methyl-2-{[4-(methylamino)-9,10-dioxo-9,10-dihydroanthracen-1-yl]amino}benzene-1-sulfonic acid sodium
Synonyms: 5-methyl-2-{[4-(methylamino)-9,10-dioxoanthracen-1-yl]amino}benzenesulfonic acid sodium
Molecular Weight (g/mol): 445.44
Molecular Formula: C22H18N2NaO5S
InChi Key: NUFVZFLCRLMJCW-UHFFFAOYSA-N
SMILES: [Na].CNC1=CC=C(NC2=CC=C(C)C=C2S(O)(=O)=O)C2=C1C(=O)C1=CC=CC=C1C2=O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results