accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 64115-88-4

FFFOBr

CAS RN 64115-88-4

IUPAC Name: 1-bromo-2-(trifluoromethoxy)benzene
Synonyms: 1-bromo-2-(trifluoromethoxy)benzene
Molecular Weight (g/mol): 241.01
Molecular Formula: C7H4BrF3O
InChi Key: ITYCJCVRPBLODP-UHFFFAOYSA-N
SMILES: FC(F)(F)OC1=CC=CC=C1Br
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results