accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 64118-84-9

IUPAC Name: 2-{2-[(2,6-dichloro-4-hydroxyphenyl)amino]phenyl}acetic acid
Synonyms: 4'-hydroxydiclofenac 4'-OH dcf 4'-hydroxy diclofenac
Molecular Weight (g/mol): 312.15
Molecular Formula: C14H11Cl2NO3
InChi Key: KGVXVPRLBMWZLG-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CC=CC=C1NC1=C(Cl)C=C(O)C=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results