accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOF

CAS RN 64123-77-9

IUPAC Name: methyl 2-(3-fluorophenyl)acetate
Synonyms: methyl 2-(3-fluorophenyl)acetate
Molecular Weight (g/mol): 168.17
Molecular Formula: C9H9FO2
InChi Key: OXYLCGRXCQARQV-UHFFFAOYSA-N
SMILES: COC(=O)CC1=CC=CC(F)=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)