accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3NNH2H2NNOOHO

CAS RN 641569-96-2

IUPAC Name: ethyl 3-[(diaminomethylidene)amino]-4-methylbenzoate; nitric acid
Synonyms: ethyl 3-[(diaminomethylidene)amino]-4-methylbenzoate; nitric acid
Molecular Weight (g/mol): 284.27
Molecular Formula: C11H16N4O5
InChi Key: YQMZYKJPGMVZJL-UHFFFAOYSA-N
SMILES: O[N+]([O-])=O.CCOC(=O)C1=CC(N=C(N)N)=C(C)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results