accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH2FMgCl

CAS RN 64168-34-9

IUPAC Name: magnesium(2+) (3-fluorophenyl)methanide chloride
Synonyms: magnesium(2+) (3-fluorophenyl)methanide chloride
Molecular Weight (g/mol): 168.88
Molecular Formula: C7H6ClFMg
InChi Key: KZGFEBLGGYOPML-UHFFFAOYSA-M
SMILES: [Mg++].[Cl-].[CH2-]C1=CC(F)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results