accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 64183-27-3

IUPAC Name: (2R,3R,4R,5R)-5-(6-amino-9H-purin-9-yl)-4-fluoro-2-(hydroxymethyl)oxolan-3-ol
Synonyms: (2R,3R,4R,5R)-5-(6-aminopurin-9-yl)-4-fluoro-2-(hydroxymethyl)oxolan-3-ol
Molecular Weight (g/mol): 269.24
Molecular Formula: C10H12FN5O3
InChi Key: ZGYYPTJWJBEXBC-GPGUAWMRNA-N
SMILES: NC1=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3F)C2=NC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results