accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOOOOOH

CAS RN 64187-47-9

IUPAC Name: 1-[(benzyloxy)carbonyl]-4-oxopyrrolidine-2-carboxylic acid
Synonyms: 1-[(benzyloxy)carbonyl]-4-oxopyrrolidine-2-carboxylic acid
Molecular Weight (g/mol): 263.25
Molecular Formula: C13H13NO5
InChi Key: RPLLCMZOIFOBIF-UHFFFAOYNA-N
SMILES: OC(=O)C1CC(=O)CN1C(=O)OCC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)