accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6419-19-8

IUPAC Name: hexapotassium {[bis(phosphonatomethyl)amino]methyl}phosphonate
Synonyms: aminotrimethylene phosphonic acid
Molecular Weight (g/mol): 527.59
Molecular Formula: C3H6K6NO9P3
InChi Key: DKPHLYCEFBDQKM-UHFFFAOYSA-H
SMILES: [K+].[K+].[K+].[K+].[K+].[K+].[O-]P([O-])(=O)CN(CP([O-])([O-])=O)CP([O-])([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results