CAS RN 64198-16-9
IUPAC Name:
tetramethyl-4,9-dioxatricyclo[5.3.0.0²,⁶]decane-3,5,8,10-tetrone
Synonyms:
tetramethyl-4,9-dioxatricyclo[5.3.0.0²,⁶]decane-3,5,8,10-tetrone
Molecular Weight (g/mol):
252.22
Molecular Formula:
C12H12O6
InChi Key:
GTDPSWPPOUPBNX-UHFFFAOYSA-N
SMILES:
CC12C(=O)OC(=O)C1(C)C1(C)C(=O)OC(=O)C21C
1
–
1
of
1
results
1
1,2,3,4-Tetramethyl-1,2,3,4-cyclobutanetetracarboxylic Dianhydride, TCI America™
CAS: 64198-16-9 Molecular Formula: C12H12O6 Molecular Weight (g/mol): 252.22 InChI Key: GTDPSWPPOUPBNX-UHFFFAOYSA-N