accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOH3CCH3H3CH3COO(E/Z)

CAS RN 642066-70-4

IUPAC Name: 4,4,5,5-tetramethyl-2-[3-(oxan-2-yloxy)prop-1-en-1-yl]-1,3,2-dioxaborolane
Synonyms: 4,4,5,5-tetramethyl-2-[3-(oxan-2-yloxy)prop-1-en-1-yl]-1,3,2-dioxaborolane
Molecular Weight (g/mol): 268.16
Molecular Formula: C14H25BO4
InChi Key: MHSOBXCZCRNELG-UHFFFAOYNA-N
SMILES: CC1(C)OB(OC1(C)C)C=CCOC1CCCCO1
Quantity 
  • (2)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)