accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6424-15-3

IUPAC Name: bis(2-hydroxypropane-1,2,3-tricarboxylic acid) tricobalt dihydrate
Synonyms: bis(citric acid) tricobalt dihydrate
Molecular Weight (g/mol): 597.08
Molecular Formula: C12H20Co3O16
InChi Key: SKMIIAIULKZNRQ-UHFFFAOYSA-N
SMILES: O.O.[Co].[Co].[Co].OC(=O)CC(O)(CC(O)=O)C(O)=O.OC(=O)CC(O)(CC(O)=O)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results