accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 64276-62-6

IUPAC Name: ethyl 4-phenyl-1H-pyrrole-3-carboxylate
Synonyms: ethyl 4-phenyl-1H-pyrrole-3-carboxylate
Molecular Weight (g/mol): 215.25
Molecular Formula: C13H13NO2
InChi Key: VNZGRYAVCWGZCT-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CNC=C1C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results