accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOHNOO

CAS RN 642995-15-1

IUPAC Name: 2-{1,3-dioxo-1H,2H,3H-benzo[f]isoindol-2-yl}cyclohexane-1-carboxylic acid
Synonyms: 2-{1,3-dioxobenzo[f]isoindol-2-yl}cyclohexane-1-carboxylic acid
Molecular Weight (g/mol): 323.35
Molecular Formula: C19H17NO4
InChi Key: RPVXICJCKYVHHZ-UHFFFAOYNA-N
SMILES: OC(=O)C1CCCCC1N1C(=O)C2=C(C=C3C=CC=CC3=C2)C1=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results