accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 643-43-6

IUPAC Name: 2-(2,4-dinitrophenyl)acetate
Synonyms: 2-(2,4-dinitrophenyl)acetate
Molecular Weight (g/mol): 225.14
Molecular Formula: C8H5N2O6
InChi Key: KCNISYPADDTFDO-UHFFFAOYSA-M
SMILES: [O-]C(=O)CC1=CC=C(C=C1[N+]([O-])=O)[N+]([O-])=O
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)