accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3O

CAS RN 643-65-2

IUPAC Name: (3-methylphenyl)(phenyl)methanone
Synonyms: 3-methylphenyl phenyl ketone
Molecular Weight (g/mol): 196.25
Molecular Formula: C14H12O
InChi Key: URBLVRAVOIVZFJ-UHFFFAOYSA-N
SMILES: CC1=CC=CC(=C1)C(=O)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results