accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 64325-78-6

IUPAC Name: N-{9-[(2R,4S,5R)-5-{[bis(4-methoxyphenyl)(phenyl)methoxy]methyl}-4-hydroxyoxolan-2-yl]-9H-purin-6-yl}benzamide
Synonyms: N-{9-[(2R,4S,5R)-5-{[bis(4-methoxyphenyl)(phenyl)methoxy]methyl}-4-hydroxyoxolan-2-yl]purin-6-yl}benzamide
Molecular Weight (g/mol): 657.73
Molecular Formula: C38H35N5O6
InChi Key: LPICNYATEWGYHI-WIHCDAFUSA-N
SMILES: COC1=CC=C(C=C1)C(OC[C@H]1O[C@H](C[C@@H]1O)N1C=NC2=C(NC(=O)C3=CC=CC=C3)N=CN=C12)(C1=CC=CC=C1)C1=CC=C(OC)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results