accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OH3CCH3OOH

CAS RN 64396-97-0

IUPAC Name: 2-(3-acetyl-2,2-dimethylcyclobutyl)acetic acid
Synonyms: (3-acetyl-2,2-dimethylcyclobutyl)acetic acid
Molecular Weight (g/mol): 184.24
Molecular Formula: C10H16O3
InChi Key: SIZDUQQDBXJXLQ-UHFFFAOYNA-N
SMILES: CC(=O)C1CC(CC(O)=O)C1(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results