accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 644-21-3

IUPAC Name: 1-ethyl-1-phenylhydrazine
Synonyms: hydrazine, 1-ethyl-1-phenyl-
Molecular Weight (g/mol): 136.2
Molecular Formula: C8H12N2
InChi Key: NQSIARGGPGHMCG-UHFFFAOYSA-N
SMILES: CCN(N)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results