accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2ClSOONHN

CAS RN 64415-13-0

IUPAC Name: 3-amino-4-chloro-N-(2-cyanoethyl)benzene-1-sulfonamide
Synonyms: 3-amino-4-chloro-N-(2-cyanoethyl)benzenesulfonamide
Molecular Weight (g/mol): 259.71
Molecular Formula: C9H10ClN3O2S
InChi Key: DNMYCTLTLUHRRT-UHFFFAOYSA-N
SMILES: NC1=CC(=CC=C1Cl)S(=O)(=O)NCCC#N
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results