CAS RN 64415-14-1
IUPAC Name:
4-phenyl-5-tetradecyl-1,3-thiazol-2-amine
Synonyms:
4-phenyl-5-tetradecyl-1,3-thiazol-2-amine
Molecular Weight (g/mol):
372.62
Molecular Formula:
C23H36N2S
InChi Key:
WYDPIVMKULHMBF-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCCCCC1=C(N=C(N)S1)C1=CC=CC=C1
1
–
1
of
1
results
1
2-Amino-4-phenyl-5-n-tetradecylthiazole 98%, Thermo Scientific™
CAS: 64415-14-1 Molecular Formula: C23H36N2S Molecular Weight (g/mol): 372.615 InChI Key: WYDPIVMKULHMBF-UHFFFAOYSA-N Synonym: 2-thiazolamine, 4-phenyl-5-tetradecyl,4-phenyl-5-tetradecylthiazol-2-amine,2-amino-4-phenyl-5-tetradecylthiazole,2-amino-4-phenyl-5-n-tetradecylthiazole,4-phenyl-5-tetradecyl-1,3-thiazole-2-ylamine,4-phenyl-5-tetradecyl-2-thiazolamine,2-thiazolamine,4-phenyl-5-tetradecyl,# PubChem CID: 73234 IUPAC Name: 4-phenyl-5-tetradecyl-1,3-thiazol-2-amine SMILES: CCCCCCCCCCCCCCC1=C(N=C(S1)N)C2=CC=CC=C2