accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 64424-12-0

IUPAC Name: lanthanum(3+) tris(2,4-dioxopentan-3-ide)
Synonyms: lanthanum(3+) triacetylacetonate
Molecular Weight (g/mol): 436.23
Molecular Formula: C15H21LaO6
InChi Key: UUYNRMJIFVOQCR-UHFFFAOYSA-N
SMILES: [La+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
Quantity 
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)