accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OCH3OCH3OCH3

CAS RN 6443-69-2

IUPAC Name: 1,2,3-trimethoxy-5-methylbenzene
Synonyms: 1,2,3-trimethoxy-5-methylbenzene
Molecular Weight (g/mol): 182.22
Molecular Formula: C10H14O3
InChi Key: KCIZTNZGSBSSRM-UHFFFAOYSA-N
SMILES: COC1=CC(C)=CC(OC)=C1OC
Molecular Weight (g/mol) 
  • (5)
  • (1)
Percent Purity 
  • (2)
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (5)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)