accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONOCH3(E)

CAS RN 6443-72-7

IUPAC Name: (2E)-3-(3,4-dimethoxyphenyl)prop-2-enenitrile
Synonyms: (2E)-3-(3,4-dimethoxyphenyl)prop-2-enenitrile
Molecular Weight (g/mol): 189.21
Molecular Formula: C11H11NO2
InChi Key: WGPTVEZJYRFEGT-ONEGZZNKSA-N
SMILES: COC1=CC=C(\C=C\C#N)C=C1OC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results