accessibility menu, dialog, popup

UserName

Viewing profile as user:
NClI

CAS RN 6443-90-9

IUPAC Name: iodochlorane; pyridine
Synonyms: iodine monochloride; pyridine
Molecular Weight (g/mol): 241.46
Molecular Formula: C5H5ClIN
InChi Key: SYWWBJKPSYWUBN-UHFFFAOYSA-N
SMILES: ClI.C1=CC=NC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results