accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 64438-53-5

OOOOOOCCCH3CH3CCH3H3CCH3H3CHHHSm

CAS RN 64438-53-5

IUPAC Name: samarium(3+) tris(2,4-dioxopentan-3-ide)
Synonyms: samarium(3+) triacetylacetonate
Molecular Weight (g/mol): 447.69
Molecular Formula: C15H21O6Sm
InChi Key: JTZJMDRBZZVNSP-UHFFFAOYSA-N
SMILES: [Sm+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results