accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 64443-05-6

IUPAC Name: λ¹-copper(1+) tetrakis(acetonitrile) hexafluoro-λ⁵-phosphanuide
Synonyms: tetrakis(acetonitrile)copper(I) hexafluorophosphate
Molecular Weight (g/mol): 372.72
Molecular Formula: C8H12CuF6N4P
InChi Key: GNQXUMGHBSAQBV-UHFFFAOYSA-N
SMILES: [Cu+].CC#N.CC#N.CC#N.CC#N.F[P-](F)(F)(F)(F)F
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results