accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 64491-68-5

IUPAC Name: (2S)-2-(methoxymethyl)oxirane
Synonyms: (2S)-2-(methoxymethyl)oxirane
Molecular Weight (g/mol): 88.11
Molecular Formula: C4H8O2
InChi Key: LKMJVFRMDSNFRT-SCSAIBSYSA-N
SMILES: COC[C@@H]1CO1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)