CAS RN 644958-86-1
CAS RN 644958-86-1
IUPAC Name:
N1,N2-bis(3,3-dimethylbutyl)-N1,N2-dimethylcyclohexane-1,2-diamine
Synonyms:
N1,N2-bis(3,3-dimethylbutyl)-N1,N2-dimethylcyclohexane-1,2-diamine
Molecular Weight (g/mol):
310.57
Molecular Formula:
C20H42N2
InChi Key:
VGCWVKVNKNXOGZ-UHFFFAOYNA-N
SMILES:
CN(CCC(C)(C)C)C1CCCCC1N(C)CCC(C)(C)C
1
–
1
of
1
results
1
(1R,2R)-N,N'-Dimethyl-N,N'-bis(3,3-dimethylbutyl)cyclohexane-1,2-diamine 97.0+%, TCI America™
CAS: 644958-86-1 Molecular Formula: C20H42N2 Molecular Weight (g/mol): 310.57 InChI Key: VGCWVKVNKNXOGZ-QZTJIDSGSA-N PubChem CID: 44629980 IUPAC Name: (1R,2R)-1-N,2-N-bis(3,3-dimethylbutyl)-1-N,2-N-dimethylcyclohexane-1,2-diamine SMILES: CC(C)(C)CCN(C)C1CCCCC1N(C)CCC(C)(C)C