accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NH3CNNNNCH3H3CNCH3CH3

CAS RN 645-05-6

IUPAC Name: N2,N2,N4,N4,N6,N6-hexamethyl-1,3,5-triazine-2,4,6-triamine
Synonyms: altretamine hexalen hexamethyl melamine hexastat hexaloids altretamina altretaminum hexinawas HMM HTM
Molecular Weight (g/mol): 210.29
Molecular Formula: C9H18N6
InChi Key: UUVWYPNAQBNQJQ-UHFFFAOYSA-N
SMILES: CN(C)C1=NC(=NC(=N1)N(C)C)N(C)C
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results