accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3CH3O

CAS RN 645-13-6

IUPAC Name: 1-[4-(propan-2-yl)phenyl]ethan-1-one
Synonyms: P-isopropylacetophenone
Molecular Weight (g/mol): 162.23
Molecular Formula: C11H14O
InChi Key: PDLCCNYKIIUWHA-UHFFFAOYSA-N
SMILES: CC(C)C1=CC=C(C=C1)C(C)=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)
  • (2)
  • (3)
  • (2)