accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 645-33-0

IUPAC Name: hydrogen 5-(2-aminoethyl)-2-methoxyphenol chloride
Synonyms: hydrogen 5-(2-aminoethyl)-2-methoxyphenol chloride
Molecular Weight (g/mol): 203.67
Molecular Formula: C9H14ClNO2
InChi Key: KAAFITWSSODFMA-UHFFFAOYSA-N
SMILES: [H+].[Cl-].COC1=CC=C(CCN)C=C1O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)