accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CNHCH3H3CCH3CH3

CAS RN 645-41-0

IUPAC Name: tris(3-methylbutyl)azanium
Synonyms: tris(3-methylbutyl)azanium
Molecular Weight (g/mol): 228.44
Molecular Formula: C15H34N
InChi Key: QKVUSSUOYHTOFQ-UHFFFAOYSA-O
SMILES: CC(C)CC[NH+](CCC(C)C)CCC(C)C
Grade 
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results