accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 645-43-2

IUPAC Name: N''-[2-(azocan-1-yl)ethyl]guanidine; sulfuric acid
Synonyms: guanethidine
Molecular Weight (g/mol): 296.39
Molecular Formula: C10H24N4O4S
InChi Key: YUFWAVFNITUSHI-UHFFFAOYSA-N
SMILES: OS(O)(=O)=O.NC(N)=NCCN1CCCCCCC1
Concentration 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results