accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NSHNNH

CAS RN 645-48-7

IUPAC Name: (phenylamino)thiourea
Synonyms: phenylaminothiourea
Molecular Weight (g/mol): 167.23
Molecular Formula: C7H9N3S
InChi Key: JDXKTOBMLZLCSB-UHFFFAOYSA-N
SMILES: NC(=S)NNC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results