accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 645-54-5

IUPAC Name: 2-phenylethanethioamide
Synonyms: 2-phenylethanethioamide
Molecular Weight (g/mol): 151.23
Molecular Formula: C8H9NS
InChi Key: CJXBHFANXQMZBF-UHFFFAOYSA-N
SMILES: NC(=S)CC1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)