CAS RN 645-96-5
IUPAC Name:
phenylsulfanyl
Synonyms:
phenylsulfanyl group
Molecular Weight (g/mol):
156.08
Molecular Formula:
C6H5Se
InChi Key:
YWBHROUQJYHSOR-UHFFFAOYSA-N
SMILES:
[Se]C1=CC=CC=C1
1
–
3
of
3
results
1
Promotions Available
Phenylselenol, 90+%
Phenylselenol, 90+%
CAS: 645-96-5 Molecular Formula: C6H6Se MDL Number: MFCD00000001 Synonym: Benzeneselenol; Selenophenol
2
Benzeneselenol 95.0+%, TCI America™
CAS: 645-96-5 Molecular Formula: C6H6Se MDL Number: MFCD00000001 Synonym: Hydroselenobenzene, Phenylselenol
3
Phenylselenol, Thermo Scientific™
CAS: 645-96-5 MDL Number: MFCD00000001 Synonym: Benzeneselenol; Selenophenol