accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H2NSOOOCH3Na

CAS RN 64501-84-4

IUPAC Name: sodium 5-amino-2,4-dimethylbenzene-1-sulfonate
Synonyms: sodium 5-amino-2,4-dimethylbenzenesulfonate
Molecular Weight (g/mol): 223.22
Molecular Formula: C8H10NNaO3S
InChi Key: ZETYURDDFWHYDN-UHFFFAOYSA-M
SMILES: [Na+].CC1=CC(C)=C(C=C1N)S([O-])(=O)=O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results