accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOHHNSOONOO

CAS RN 64501-87-7

IUPAC Name: 2-(4-nitrobenzenesulfonamido)-3-phenylpropanoic acid
Synonyms: 2-(4-nitrobenzenesulfonamido)-3-phenylpropanoic acid
Molecular Weight (g/mol): 350.35
Molecular Formula: C15H14N2O6S
InChi Key: JVHRPLXWZFPXIJ-UHFFFAOYNA-N
SMILES: OC(=O)C(CC1=CC=CC=C1)NS(=O)(=O)C1=CC=C(C=C1)[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results