CAS RN 6459-94-5
CAS RN 6459-94-5
IUPAC Name:
disodium (8E)-8-{2-[3,3'-dimethyl-4'-(2-{4-[(4-methylbenzenesulfonyl)oxy]phenyl}diazen-1-yl)-[1,1'-biphenyl]-4-yl]hydrazin-1-ylidene}-7-oxo-7,8-dihydronaphthalene-1,3-disulfonate
Synonyms:
disodium (8E)-8-{2-[3,3'-dimethyl-4'-(2-{4-[(4-methylbenzenesulfonyl)oxy]phenyl}diazen-1-yl)-[1,1'-biphenyl]-4-yl]hydrazin-1-ylidene}-7-oxonaphthalene-1,3-disulfonate
Molecular Weight (g/mol):
830.81
Molecular Formula:
C37H28N4Na2O10S3
InChi Key:
PVLKQJLDEXCKNY-JYLPYTDFSA-L
SMILES:
[Na+].[Na+].CC1=CC=C(C=C1)S(=O)(=O)OC1=CC=C(C=C1)N=NC1=CC=C(C=C1C)C1=CC=C(N\N=C2\C(=O)C=CC3=CC(=CC(=C23)S([O-])(=O)=O)S([O-])(=O)=O)C(C)=C1
1
–
1
of
1
results
1
Acid Red 114, TCI America™

CAS: 6459-94-5 Molecular Formula: C37H28N4Na2O10S3 Molecular Weight (g/mol): 830.809 MDL Number: MFCD00003894 InChI Key: PVLKQJLDEXCKNY-UHFFFAOYSA-L PubChem CID: 5479172 IUPAC Name: disodium;8-[[2-methyl-4-[3-methyl-4-[[4-(4-methylphenyl)sulfonyloxyphenyl]diazenyl]phenyl]phenyl]hydrazinylidene]-7-oxonaphthalene-1,3-disulfonate SMILES: CC1=CC=C(C=C1)S(=O)(=O)OC2=CC=C(C=C2)N=NC3=C(C=C(C=C3)C4=CC(=C(C=C4)NN=C5C(=O)C=CC6=CC(=CC(=C65)S(=O)(=O)[O-])S(=O)(=O)[O-])C)C.[Na+].[Na+]