accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSOHO

CAS RN 646-01-5

IUPAC Name: 3-(methylsulfanyl)propanoic acid
Synonyms: 3-methylthiopropionic acid 3-methyl-thiopropionic acid
Molecular Weight (g/mol): 120.17
Molecular Formula: C4H8O2S
InChi Key: CAOMCZAIALVUPA-UHFFFAOYSA-N
SMILES: CSCCC(O)=O
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)