accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 646-24-2

IUPAC Name: nonane-1,9-bis(aminium)
Synonyms: nonane-1,9-bis(aminium)
Molecular Weight (g/mol): 160.3
Molecular Formula: C9H24N2
InChi Key: SXJVFQLYZSNZBT-UHFFFAOYSA-P
SMILES: [NH3+]CCCCCCCCC[NH3+]
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)