accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 646-25-3

IUPAC Name: decane-1,10-diamine
Synonyms: 1,10-diaminodecane
Molecular Weight (g/mol): 172.32
Molecular Formula: C10H24N2
InChi Key: YQLZOAVZWJBZSY-UHFFFAOYSA-N
SMILES: NCCCCCCCCCCN
Boiling Point 
  • (2)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (3)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (4)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)