accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNNH2HCl

CAS RN 646051-17-4

IUPAC Name: 2-(4-methylpiperazin-1-yl)ethan-1-amine hydrochloride
Synonyms: 2-(4-methylpiperazin-1-yl)ethanamine hydrochloride
Molecular Weight (g/mol): 179.69
Molecular Formula: C7H18ClN3
InChi Key: KGJPTLVFRDJLHM-UHFFFAOYSA-N
SMILES: Cl.CN1CCN(CCN)CC1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results