accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClSOOSClOO

CAS RN 6461-76-3

IUPAC Name: benzene-1,2-disulfonyl dichloride
Synonyms: benzene-1,2-disulfonyl dichloride
Molecular Weight (g/mol): 275.11
Molecular Formula: C6H4Cl2O4S2
InChi Key: YBGQXNZTVFEKEN-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)C1=CC=CC=C1S(Cl)(=O)=O
Quantity 
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)