CAS RN 64657-18-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Forskolin, 1,9-Dideoxy-, Coleus forskohlii
Natural product that does not stimulate adenylate cyclase.