accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SnCH3CH3OH3C(E/Z)

CAS RN 64724-29-4

IUPAC Name: tributyl(2-ethoxyethenyl)stannane
Synonyms: tributyl(2-ethoxyethenyl)stannane
Molecular Weight (g/mol): 361.16
Molecular Formula: C16H34OSn
InChi Key: WARKYKQCOXTIAO-UHFFFAOYSA-N
SMILES: CCCC[Sn](CCCC)(CCCC)C=COCC
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)