accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaNaNaH2NNNNHNNHNHNOSOOONNNH2NH2SOOOOSOOONH2(E/Z)(Z)(Z)(E/Z)

CAS RN 6473-13-8

IUPAC Name: trisodium (3Z)-6-[2-(2,4-diaminophenyl)diazen-1-yl]-3-(2-{4-[(4-{2-[(2Z)-7-[2-(2,4-diaminophenyl)diazen-1-yl]-1-oxo-3-sulfonato-1,2-dihydronaphthalen-2-ylidene]hydrazin-1-yl}-2-sulfonatophenyl)amino]phenyl}hydrazin-1-ylidene)-4-oxo-3,4-dihydronaphthalene-2-sulfonate
Synonyms: trisodium (3Z)-6-[2-(2,4-diaminophenyl)diazen-1-yl]-3-(2-{4-[(4-{2-[(2Z)-7-[2-(2,4-diaminophenyl)diazen-1-yl]-1-oxo-3-sulfonatonaphthalen-2-ylidene]hydrazin-1-yl}-2-sulfonatophenyl)amino]phenyl}hydrazin-1-ylidene)-4-oxonaphthalene-2-sulfonate
Molecular Weight (g/mol): 1083.97
Molecular Formula: C44H32N13Na3O11S3
InChi Key: KCLJSZPAUJRRGH-FIDRBWEZSA-K
SMILES: [Na+].[Na+].[Na+].NC1=CC=C(N=NC2=CC=C3C=C(\C(=N/NC4=CC=C(NC5=CC=C(N\N=C6\C(=O)C7=CC(=CC=C7C=C6S([O-])(=O)=O)N=NC6=CC=C(N)C=C6N)C=C5S([O-])(=O)=O)C=C4)C(=O)C3=C2)S([O-])(=O)=O)C(N)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results