accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNH3CH3CCH3CH3O

CAS RN 647824-51-9

IUPAC Name: 1-tert-butyl-3,5-dimethyl-1H-pyrazole-4-carbaldehyde
Synonyms: 1-tert-butyl-3,5-dimethylpyrazole-4-carbaldehyde
Molecular Weight (g/mol): 180.25
Molecular Formula: C10H16N2O
InChi Key: WQHXOHHEPNBXMF-UHFFFAOYSA-N
SMILES: CC1=NN(C(C)=C1C=O)C(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results